Assay Details
| Gene Symbol: C330022C24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045717.1 | 2 - 3 | 410 | 65 | ||
| GenBank mRNA | AK021219.1 | - | 2 - 3 | 411 | 65 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | urate (5-hydroxyiso-) hydrolase | Urah |
1190003J15Rik, 2810420C16Rik |
-3.2kb | Anti-sense | Chr. 7 - 140835023 - 140837968 on NCBI Build GRCm38 |
| Downstream |
secretoglobin, family 1C, member 1 |
Scgb1c1 | Ryd5 | 3.6kb | Anti-sense | Chr. 7 - 140845565 - 140846769 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

C330022C24Rik
Genomic Map