Assay Details
Gene Symbol: Ccnd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK020317.1 | - | 1 - 2 | 52 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
