Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200029364.mm.1 |
NM_001159548.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029364.mm.1 |
NM_001159548.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029364.mm.1 |
NM_001159549.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029364.mm.1 |
NM_001159549.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029364.mm.1 |
NM_010264.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029364.mm.1 |
NM_010264.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029364.mm.1 |
TC0200003424.mm.1 |
MTA TC |