Assay Details
| Gene Symbol: Hat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026115.4 | NP_080391.2 | 6 - 7 | 630 | 139 | |
| GenBank mRNA | AK010715.1 | - | 6 - 7 | 639 | 140 | |
| AK041700.1 | - | 6 - 7 | 634 | 139 | ||
| AK137648.1 | - | 6 - 7 | 648 | 139 | ||
| AK169315.1 | - | 6 - 7 | 633 | 139 | ||
| BC038262.1 | - | 4 - 5 | 510 | 139 | ||
| BC055460.1 | - | 5 - 6 | 597 | 4457298 | 139 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hat1
Genomic Map