Assay Details
Gene Symbol: Ext2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010163.3 | NP_034293.2 | 11 - 12 | 2010 | 106 | |
GenBank mRNA | AK134438.1 | - | 7 - 8 | 951 | 106 | |
AK148090.1 | - | 11 - 12 | 2000 | 106 | ||
AK148369.1 | - | 10 - 11 | 1771 | 106 | ||
AK150155.1 | - | 11 - 12 | 2002 | 106 | ||
AK164342.1 | - | 11 - 12 | 1996 | 106 | ||
AK184805.1 | - | 1 - 2 | 122 | 106 | ||
AK188211.1 | - | 1 - 2 | 94 | 106 | ||
BC006597.1 | - | 11 - 12 | 1983 | 3965079 | 106 | |
U67837.1 | - | 11 - 12 | 1985 | 106 | ||
U72141.1 | - | 11 - 12 | 1989 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
