Assay Details
| Gene Symbol: Ext2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010163.3 | NP_034293.2 | 12 - 13 | 2147 | 68 | |
| GenBank mRNA | AK134438.1 | - | 8 - 9 | 1088 | 68 | |
| AK148090.1 | - | 12 - 13 | 2137 | 68 | ||
| AK148369.1 | - | 11 - 12 | 1908 | 68 | ||
| AK150155.1 | - | 12 - 13 | 2139 | 68 | ||
| AK164342.1 | - | 12 - 13 | 2133 | 68 | ||
| AK184805.1 | - | 2 - 3 | 259 | 68 | ||
| AK188211.1 | - | 2 - 3 | 231 | 68 | ||
| BC006597.1 | - | 12 - 13 | 2120 | 3965079 | 68 | |
| U67837.1 | - | 12 - 13 | 2122 | 68 | ||
| U72141.1 | - | 12 - 13 | 2126 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ext2
Genomic Map