Assay Details
Gene Symbol: Ext2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010163.3 | NP_034293.2 | 6 - 7 | 1296 | 117 | |
GenBank mRNA | AK082572.1 | - | 6 - 7 | 1278 | 117 | |
AK134438.1 | - | 2 - 3 | 237 | 117 | ||
AK148090.1 | - | 6 - 7 | 1286 | 117 | ||
AK148369.1 | - | 5 - 6 | 1057 | 117 | ||
AK150155.1 | - | 6 - 7 | 1288 | 117 | ||
AK152265.1 | - | 1396 | 117 | |||
AK152840.1 | - | 1398 | 117 | |||
AK164342.1 | - | 6 - 7 | 1282 | 117 | ||
AK187641.1 | - | 1 - 2 | 99 | 117 | ||
BC006597.1 | - | 6 - 7 | 1269 | 3965079 | 117 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
