Assay Details
| Gene Symbol: Syt9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021889.4 | NP_068689.2 | 3 - 4 | 1258 | 75 | |
| GenBank mRNA | AB026802.1 | - | 3 - 4 | 1057 | 75 | |
| AK089115.1 | - | 3 - 4 | 1259 | 75 | ||
| AK156631.1 | - | 3 - 4 | 1259 | 75 | ||
| BC132495.1 | - | 3 - 4 | 1245 | 40130772 | 75 | |
| BC137904.1 | - | 3 - 4 | 1245 | 8860923 | 75 | |
| BC145024.1 | - | 3 - 4 | 1245 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Syt9
Genomic Map