Assay Details
| Gene Symbol: Mcm8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291054.1 | NP_001277983.1 | 1 - 2 | 327 | 72 | |
| NM_025676.4 | NP_079952.2 | 1 - 2 | 327 | 72 | ||
| GenBank mRNA | AK012845.1 | - | 1 - 2 | 286 | 72 | |
| AK017607.1 | - | 1 - 2 | 225 | 72 | ||
| AK019268.1 | - | 1 - 2 | 286 | 72 | ||
| AK133858.1 | - | 1 - 2 | 284 | 72 | ||
| BC046780.1 | - | 1 - 2 | 274 | 5716136 | 72 | |
| BC052070.1 | - | 1 - 2 | 133 | 5716125 | 72 | |
| BY766288.1 | - | 1 - 2 | 327 | 72 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK012845.1, AK019268.1, AK133858.1, BC046780.1, BC052070.1, NM_001291054.1, NM_025676.4 transcripts.
Set Membership: |
|

Mcm8
Genomic Map