Assay Details
| Gene Symbol: Mcm8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291054.1 | NP_001277983.1 | 4 - 5 | 662 | 148 | |
| NM_025676.4 | NP_079952.2 | 3 - 4 | 578 | 148 | ||
| GenBank mRNA | AK017607.1 | - | 4 - 5 | 560 | 148 | |
| AK133858.1 | - | 4 - 5 | 619 | 148 | ||
| BC046780.1 | - | 4 - 5 | 609 | 5716136 | 148 | |
| BC052070.1 | - | 3 - 4 | 384 | 5716125 | 148 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mcm8
Genomic Map