Assay Details
| Gene Symbol: Mcm8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291054.1 | NP_001277983.1 | 6 - 7 | 910 | 70 | |
| NM_025676.4 | NP_079952.2 | 5 - 6 | 826 | 70 | ||
| GenBank mRNA | AK017607.1 | - | 6 - 7 | 808 | 70 | |
| AK133858.1 | - | 6 - 7 | 867 | 70 | ||
| BC046780.1 | - | 6 - 7 | 857 | 5716136 | 70 | |
| BC052070.1 | - | 5 - 6 | 632 | 5716125 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mcm8
Genomic Map