Assay Details
| Gene Symbol: Zkscan2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081329.1 | NP_001074798.1 | 6 - 7 | 2437 | 93 | |
| GenBank mRNA | AK034954.1 | - | 6 - 7 | 2439 | 93 | |
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1992227 | BC054371.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1992227 | BC028272.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1992227 | NM_001081329.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 54618 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 391177 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 563295 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 851615 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 156039 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 355066 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 801044 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 860020 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 872466 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 597589 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 23855 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 443019 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 11857 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 117339 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 98160 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 86273 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 17947 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P154426 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P109843 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P157546 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P147613 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P106362 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1992227 | BC054371.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1992227 | BC028272.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1992227 | NM_001081329.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P109843 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06352162 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P154426 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P157546 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P147613 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P106362 | - | - |
| Whole Mouse Genome G4122A | A_52_P160527 | BC054371.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P160527 | BC028272.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P103298 | BC054371.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P103298 | BC028272.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P103298 | NM_001081329.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P103298 | AK034954.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P147613 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P109843 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P154426 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P106362 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P157546 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06175772 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06175773 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06175776 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06175774 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06175782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484697 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25484735 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06440718 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484809 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484791 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484732 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484639 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06440681 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484738 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484658 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484724 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484654 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484644 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484660 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484778 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484786 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484785 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484787 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484770 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484740 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484808 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484656 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484748 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484723 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06650137 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484736 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484754 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484762 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484651 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484790 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484804 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484647 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484659 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484646 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484758 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484661 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484782 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484796 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484743 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484777 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484805 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484742 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06118081 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484731 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484635 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06440691 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484637 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484780 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484766 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484793 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06011979 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06157178 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484801 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06150059 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06440710 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06495859 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484642 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484764 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484792 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484811 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484652 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484799 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06297970 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06221600 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484725 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484772 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484788 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484726 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484717 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484729 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484773 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484746 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484744 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484722 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484757 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484728 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484803 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484718 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484720 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484813 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484814 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484749 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25484781 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10567732 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10567731 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10567729 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10567730 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10567726 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10567734 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10567733 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10567727 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10567728 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10567725 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 106840341 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 105050039 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 106840341 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2697712 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2697712 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1254594 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1223952 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 105050039 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 106840341 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 105050039 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106840341 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2873274 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2873274 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2563870 | - | - |

Zkscan2
Genomic Map