Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700015154.mm.1 |
TC0700001773.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700015154.mm.1 |
NM_025304.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015154.mm.1 |
NM_025304.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015152.mm.1 |
TC0700001773.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700015152.mm.1 |
NM_025304.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015152.mm.1 |
NM_025304.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015151.mm.1 |
TC0700001773.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700015151.mm.1 |
NM_025304.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015151.mm.1 |
NM_025304.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015153.mm.1 |
TC0700001773.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700015153.mm.1 |
NM_025304.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015153.mm.1 |
NM_025304.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015155.mm.1 |
TC0700001773.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700015155.mm.1 |
NM_025304.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015155.mm.1 |
NM_025304.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015150.mm.1 |
TC0700001773.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700015150.mm.1 |
NM_025304.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015150.mm.1 |
NM_025304.3 |
Refseq |