Assay Details
| Gene Symbol: P2rx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145526.2 | NP_663501.2 | 6 - 7 | 1099 | 64 | |
| GenBank mRNA | AK140283.1 | - | 6 - 7 | 2367 | 64 | |
| AK141847.1 | - | 6 - 7 | 764 | 64 | ||
| BC023089.1 | - | 6 - 7 | 752 | 3984739 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

P2rx3
Genomic Map