Assay Details
| Gene Symbol: Nsmce1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026330.3 | NP_080606.2 | 6 - 7 | 744 | 82 | |
| GenBank mRNA | AK009715.1 | - | 6 - 7 | 633 | 82 | |
| AK160217.1 | - | 6 - 7 | 644 | 82 | ||
| AK166410.1 | - | 6 - 7 | 82 | |||
| BC031848.1 | - | 6 - 7 | 3984224 | 82 | ||
| BC049558.1 | - | 6 - 7 | 737 | 6707952 | 82 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Nsmce1
Genomic Map