Assay Details
| Gene Symbol: Eif3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146200.1 | NP_666312.1 | 11 - 12 | 1241 | 65 | |
| GenBank mRNA | AK088076.1 | - | 11 - 12 | 1284 | 65 | |
| AK152693.1 | - | 11 - 12 | 1259 | 65 | ||
| AK160078.1 | - | 11 - 12 | 1261 | 65 | ||
| AK168160.1 | - | 11 - 12 | 1245 | 65 | ||
| AK168174.1 | - | 11 - 12 | 1245 | 65 | ||
| BC024855.1 | - | 11 - 12 | 1241 | 5359060 | 65 | |
| BC025032.1 | - | 11 - 12 | 1262 | 4947435 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Eif3c
Genomic Map