Assay Details
Gene Symbol: Mapk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011952.2 | NP_036082.1 | 3 - 4 | 561 | 63 | |
GenBank mRNA | AK155287.1 | - | 2 - 3 | 503 | 63 | |
BC013754.1 | - | 3 - 4 | 394 | 63 | ||
BC029712.1 | - | 3 - 4 | 555 | 5041517 | 63 | |
BQ929318.1 | - | 3 - 4 | 561 | 63 | ||
Z14249.1 | - | 3 - 4 | 532 | 63 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
