Assay Details
Gene Symbol: Angptl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020581.2 | NP_065606.2 | 2 - 3 | 623 | 108 | |
GenBank mRNA | AB054540.1 | - | 2 - 3 | 509 | 108 | |
AF123261.1 | - | 2 - 3 | 554 | 108 | ||
AF169313.1 | - | 2 - 3 | 486 | 108 | ||
AF278699.1 | - | 2 - 3 | 558 | 108 | ||
AK009161.1 | - | 2 - 3 | 329 | 108 | ||
AK014564.1 | - | 624 | 108 | |||
AK032491.1 | - | 2 - 3 | 592 | 108 | ||
AK132761.1 | - | 2 - 3 | 592 | 108 | ||
BC006611.1 | - | 2 - 3 | 575 | 3966729 | 108 | |
BC021343.1 | - | 2 - 3 | 589 | 4194173 | 108 | |
BC025797.1 | - | 2 - 3 | 559 | 5137159 | 108 | |
KJ650202.1 | - | 2 - 3 | 440 | 108 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
