Assay Details
Gene Symbol: Angptl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020581.2 | NP_065606.2 | 5 - 6 | 956 | 80 | |
GenBank mRNA | AB054540.1 | - | 5 - 6 | 842 | 80 | |
AF169313.1 | - | 5 - 6 | 819 | 80 | ||
AK009161.1 | - | 5 - 6 | 662 | 80 | ||
AK014564.1 | - | 957 | 80 | |||
AK032491.1 | - | 5 - 6 | 924 | 80 | ||
AK132761.1 | - | 5 - 6 | 925 | 80 | ||
AK134419.1 | - | 5 - 6 | 921 | 80 | ||
BC006611.1 | - | 5 - 6 | 908 | 3966729 | 80 | |
BC021343.1 | - | 5 - 6 | 922 | 4194173 | 80 | |
BC025797.1 | - | 5 - 6 | 892 | 5137159 | 80 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
