Assay Details
Gene Symbol: Daxx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007829.4 | NP_031855.3 | 1 - 2 | 151 | 131 | |
GenBank mRNA | AK090356.1 | - | 1 - 2 | 82 | 131 | |
AK133863.1 | - | 1 - 2 | 59 | 131 | ||
AK144332.1 | - | 1 - 2 | 82 | 131 | ||
AK145906.1 | - | 1 - 2 | 82 | 131 | ||
AK146743.1 | - | 1 - 2 | 78 | 131 | ||
AK151279.1 | - | 1 - 2 | 71 | 131 | ||
DV653043.1 | - | 1 - 2 | 98 | 131 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the AK133863.1,AK145906.1,AK146743.1,NM_007829.4 transcripts.
This assay is in the 5'UTR region of the AK133863.1,AK145906.1,AK146743.1,NM_007829.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
