Assay Details
| Gene Symbol: Syce1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143765.1 | NP_001137237.1 | 3 - 4 | 236 | 81 | |
| GenBank mRNA | AK016694.1 | - | 3 - 4 | 236 | 81 | |
| BY081845.1 | - | 3 - 4 | 237 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Syce1
Genomic Map