Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700018538.mm.1 |
TC0700002065.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700018538.mm.1 |
NM_001009929.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018538.mm.1 |
NM_001009930.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018538.mm.1 |
NM_001276763.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018538.mm.1 |
NM_029426.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018538.mm.1 |
NM_029426.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018544.mm.1 |
TC0700002065.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700018544.mm.1 |
NM_001009929.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018544.mm.1 |
NM_001009930.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018544.mm.1 |
NM_001276763.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018544.mm.1 |
NM_029426.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018544.mm.1 |
NM_029426.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018541.mm.1 |
TC0700002065.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700018541.mm.1 |
NM_001009929.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018541.mm.1 |
NM_001009930.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018541.mm.1 |
NM_001276763.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018541.mm.1 |
NM_029426.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018541.mm.1 |
NM_029426.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018542.mm.1 |
TC0700002065.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700018542.mm.1 |
NM_001009929.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018542.mm.1 |
NM_001009930.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018542.mm.1 |
NM_001276763.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018542.mm.1 |
NM_029426.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018542.mm.1 |
NM_029426.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018536.mm.1 |
TC0700002065.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700018536.mm.1 |
NM_001009929.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018536.mm.1 |
NM_001009930.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018536.mm.1 |
NM_001276763.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018536.mm.1 |
NM_029426.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018536.mm.1 |
NM_029426.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018540.mm.1 |
TC0700002065.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700018540.mm.1 |
NM_001009929.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018540.mm.1 |
NM_001009930.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018540.mm.1 |
NM_001276763.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018540.mm.1 |
NM_029426.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018540.mm.1 |
NM_029426.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018537.mm.1 |
TC0700002065.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700018537.mm.1 |
NM_001009929.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018537.mm.1 |
NM_001009930.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018537.mm.1 |
NM_001276763.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018537.mm.1 |
NM_029426.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018537.mm.1 |
NM_029426.2 |
Refseq |