Assay Details
| Gene Symbol: Cpxm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018867.5 | NP_061355.3 | 3 - 4 | 665 | 95 | |
| GenBank mRNA | AF017639.1 | - | 3 - 4 | 666 | 95 | |
| AK019509.1 | - | 3 - 4 | 664 | 95 | ||
| AK038874.1 | - | 3 - 4 | 548 | 95 | ||
| BC021444.1 | - | 3 - 4 | 587 | 5038528 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cpxm2
Genomic Map