Assay Details
Gene Symbol: Dll1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007865.3 | NP_031891.2 | 9 - 10 | 2628 | 113 | |
GenBank mRNA | BC057400.1 | - | 9 - 10 | 2141 | 6402691 | 113 |
BC065063.1 | - | 9 - 10 | 2392 | 6416191 | 113 | |
X80903.1 | - | 9 - 10 | 2058 | 113 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
