Assay Details
| Gene Symbol: Mss51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029104.1 | NP_083380.1 | 4 - 5 | 790 | 77 | |
| GenBank mRNA | AK014794.1 | - | 4 - 5 | 571 | 77 | |
| BC119773.1 | - | 4 - 5 | 683 | 40107223 | 77 | |
| BC119816.1 | - | 4 - 5 | 683 | 40107221 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mss51
Genomic Map