Assay Details
| Gene Symbol: Pla2g4d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024137.1 | NP_001019308.1 | 19 - 20 | 2283 | 74 | |
| GenBank mRNA | AB195276.1 | - | 19 - 20 | 2283 | 74 | |
| AK019180.1 | - | 1 - 2 | 130 | 74 | ||
| AK145985.1 | - | 19 - 20 | 2284 | 74 | ||
| AK167492.1 | - | 19 - 20 | 2251 | 74 | ||
| BC113770.1 | - | 19 - 20 | 2280 | 40060146 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1987775 | AK167492.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1987775 | AK019180.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1987775 | AK145985.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1987775 | NM_001024137.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1987775 | AB195276.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 123694 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 260429 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 239058 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 957942 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 407438 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 194143 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 382369 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 328622 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 33312 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 170736 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 246381 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 914174 | AK003469.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 914174 | AK004704.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 914174 | AK028668.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 914174 | AK028814.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 914174 | AK133747.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 914174 | BC006578.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 914174 | NM_026176.3 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 87312 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 913241 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 212743 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 149352 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 111448 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 24890 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P156484 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1987775 | AK167492.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1987775 | AK019180.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1987775 | AK145985.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1987775 | NM_001024137.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1987775 | AB195276.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21574407 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21574400 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21574405 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21574397 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21574410 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21574381 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21574376 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21574385 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21574390 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21574384 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21574383 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21574386 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21574415 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21574417 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21574388 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21574394 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21574399 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21574413 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21574403 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21574409 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21574379 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21574401 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21574412 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21574392 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21574378 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04667417 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P156484 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04667452 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P156484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21574263 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21574385 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21574383 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21574433 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21574292 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21574426 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21574427 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21574386 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02091123 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02129899 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21574403 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02129901 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21574290 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21574354 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21574352 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01234805 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21574351 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21574384 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21574421 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21574289 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21574242 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21574349 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21574378 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21574411 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01213985 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02127417 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21574250 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21574359 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01213976 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21574413 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21574422 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21574406 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21574366 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21574240 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01213974 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02149916 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21574415 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02162296 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21574390 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21574371 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21574245 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21574409 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21574375 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21574381 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21574397 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21574355 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21574400 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21574369 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21574388 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21574370 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21574394 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21574368 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02162298 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21574423 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21574430 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01403969 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01205503 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02129904 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21574432 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01213968 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01213973 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21574249 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01476624 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21574248 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02149919 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10486427 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486428 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486432 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486436 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486444 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486442 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486431 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486443 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486438 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486426 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486447 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486441 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486446 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486433 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486445 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486437 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486435 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486439 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486430 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486440 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486434 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10486425 | - | - |
| Mouse Genome 430 2.0 Array | 1453405_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1453405_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 78390_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 78390_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200037357.mm.1 | NM_001024137.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200037357.mm.1 | NM_001024137.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200037357.mm.1 | TC0200004456.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200037358.mm.1 | NM_001024137.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200037358.mm.1 | NM_001024137.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200037358.mm.1 | TC0200004456.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1453405_PM_at | - | - |
| Mouse Expression Set 430 | 1453405_at | - | - |
| Murine Genome U74v2 Set | 167800_at | - | - |
| Murine Genome U74v2 Set | 167800_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 386604 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2700860 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2700860 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3156124 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3077168 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3077168 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3156124 | - | - |

Pla2g4d
Genomic Map