Assay Details
Gene Symbol: Olfr649 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_147055.1 | NP_667266.1 | 1 | 133 | 97 | |
GenBank mRNA | BC104080.2 | - | 1 | 126 | 97 | |
BC104081.2 | - | 1 | 126 | 97 | ||
BC119409.2 | - | 1 | 126 | 97 | ||
BC119410.2 | - | 1 | 126 | 97 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2180685 | NM_147055.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2180685 | BC104080.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2180685 | BC104081.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2180685 | BC119409.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2180685 | BC119410.2 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P123960 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2180685 | NM_147055.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2180685 | BC104080.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2180685 | BC104081.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2180685 | BC119409.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2180685 | BC119410.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P273234 | NM_147055.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P273234 | BC104080.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P273234 | BC104081.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P273234 | BC119409.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P273234 | BC119410.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25387053 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25387063 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25387047 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25387072 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25387048 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25387079 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25387084 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25387071 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25387052 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25387065 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25387067 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25387082 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25387059 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25387057 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25387050 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25387055 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25387077 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25387049 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25387062 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25387074 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25387080 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25387076 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25387069 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P123960 | - | - |
Whole Mouse Genome G4122A | A_51_P273234 | NM_147055.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P273234 | BC104080.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P273234 | BC104081.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P273234 | BC119409.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P273234 | BC119410.2 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125415 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125197 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P123960 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06172460 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06158415 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06172466 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06158414 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05885614 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25387076 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25387089 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25387082 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25387057 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25387053 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25387066 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25387050 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25387091 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25387074 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25387009 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25387049 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25387026 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25387097 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25387013 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25387058 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25387080 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25387007 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25387022 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25387063 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25387099 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25387047 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25387048 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25387105 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25387030 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25387092 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25387029 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25387102 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25387087 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25387024 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25387096 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25387069 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25387016 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25387079 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25387077 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25387065 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25387004 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25387086 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25387062 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25387027 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25387011 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25387100 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25387019 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25387059 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25387085 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25387008 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25387003 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25387048 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25387097 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25387085 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25387082 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06573868 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25387095 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25387059 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25387080 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25387049 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25387066 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06172471 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25387076 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25387050 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25387079 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25387092 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25387062 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25387052 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25387096 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25387077 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25387065 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06114142 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25387087 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06158416 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25387089 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25387053 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25387074 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25387071 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25387064 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25387072 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25387057 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25387063 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10566325 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10566324 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 316006 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 4570278 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0004570278 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2619810 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2619810 | - | - |
mouse-6 v1.1 (Illumina) | scl8860.1.1_248-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 4570278 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl8860.1.1_248-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4570278 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2619810 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2619810 | - | - |
