Assay Details
| Gene Symbol: Tsku | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024619.3 | NP_001019790.1 | 1 - 2 | 45 | 58 | |
| GenBank mRNA | AK035461.1 | - | 1 - 2 | 46 | 58 | |
| AK137208.1 | - | 1 - 2 | 46 | 58 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001024619.3 transcripts.
Set Membership: |
|

Tsku
Genomic Map