| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700012692.mm.1 |
TC0700001466.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700012692.mm.1 |
NM_147093.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700012692.mm.1 |
NM_147093.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700012693.mm.1 |
TC0700001466.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700012693.mm.1 |
NM_147093.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700012693.mm.1 |
NM_147093.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700012694.mm.1 |
TC0700001466.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700012694.mm.1 |
NM_147093.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700012694.mm.1 |
NM_147093.3 |
Refseq |