Assay Details
| Gene Symbol: Olfr1314 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146450.2 | NP_666661.2 | 1 | 69 | 63 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P629709 | NM_146450.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P629709 | NM_146450.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P174050 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P629709 | NM_146450.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P629709 | NM_146450.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P629709 | NM_146450.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P629709 | NM_146450.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P174050 | - | - |
| Whole Mouse Genome G4122A | A_52_P629709 | NM_146450.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P629709 | NM_146450.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P131840 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P174050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527320 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527298 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01484520 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527305 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02035004 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527337 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527351 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527334 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527342 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527349 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527308 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01484508 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527310 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527347 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527352 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527339 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02035006 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527324 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527346 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527325 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527299 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01484509 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527335 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527297 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527313 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527322 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527344 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01484513 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527333 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01484525 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527330 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527327 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527341 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527332 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527331 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527353 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527302 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01484534 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10485814 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10485813 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 622835 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6290551 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6290551 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006290551 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2609919 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2609919 | - | - |
| mouse-6 v1.1 (Illumina) | scl18870.1.118_4-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6290551 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl18870.1.118_4-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6290551 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2609919 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2609919 | - | - |

Olfr1314
Genomic Map