Assay Details
| Gene Symbol: Lamtor1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025605.3 | NP_079881.2 | 1 - 2 | 210 | 154 | |
| GenBank mRNA | AK010247.1 | - | 1 - 2 | 143 | 153 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_025605.3 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Lamtor1
Genomic Map