Assay Details
Gene Symbol: Olfr481 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146925.1 | NP_667136.1 | 1 | 890 | 87 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
signal transduction
G-protein coupled receptor signaling pathway
sensory perception of smell
response to stimulus
detection of chemical stimulus involved in sensory perception of smell
signal transducer activity
G-protein coupled receptor activity
olfactory receptor activity
odorant binding
signal transduction
G-protein coupled receptor signaling pathway
sensory perception of smell
response to stimulus
detection of chemical stimulus involved in sensory perception of smell
signal transducer activity
G-protein coupled receptor activity
olfactory receptor activity
odorant binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2146837 | NM_146925.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 617891 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2146837 | NM_146925.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P154115 | NM_146925.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402192 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402201 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402199 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402190 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402217 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402221 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402188 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402206 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402218 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402189 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402207 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402214 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402197 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402186 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402212 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402203 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402193 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402205 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402194 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402211 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402196 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402185 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402195 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402200 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402198 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402216 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402191 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25402208 | - | - |
Whole Mouse Genome G4122A | A_51_P154115 | NM_146925.1 | mouse_transcriptome |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402226 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402229 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402185 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402197 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402237 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402222 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402242 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402248 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402186 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402211 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402205 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402252 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402241 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402224 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402203 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402200 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402207 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402260 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402192 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402235 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402217 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402194 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402216 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402188 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402243 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402227 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402212 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402191 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402190 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402257 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402240 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402258 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402189 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402214 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402201 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402196 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402250 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402238 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402218 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402183 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402199 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402246 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402208 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402223 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402239 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402232 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402259 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402245 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25402198 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402217 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402183 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402227 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402233 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402211 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402250 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402246 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402235 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402192 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402232 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402212 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402191 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402216 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402226 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402196 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402194 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402238 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402243 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402218 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402197 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402240 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402221 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402222 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402186 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402193 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402198 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402214 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402207 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402205 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402190 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402199 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402203 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402239 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402237 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402229 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402208 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402188 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402248 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402189 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402241 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402200 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402242 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402201 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402224 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402245 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402185 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25402223 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10556130 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10556129 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 548776 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 450593 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0000450593 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2693348 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2693348 | - | - |
mouse-6 v1.1 (Illumina) | scl9357.2.1_72-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 450593 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl9357.2.1_72-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 450593 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2693348 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2693348 | - | - |
