Assay Details
| Gene Symbol: Olfr661 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146748.1 | NP_666959.1 | 1 | 851 | 116 | |
| GenBank mRNA | BC128005.1 | - | 1 | 855 | 40135960 | 116 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P519632 | NM_146748.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P519632 | AF121983.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P519632 | NM_146748.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P519632 | AF121983.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 855301 | AK016459.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 855301 | BC061039.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 855301 | NM_026624.2 | refseq_mrna_mouse_NM |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P135019 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P519632 | NM_146748.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P519632 | AF121983.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P519632 | NM_146748.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P519632 | AF121983.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P519632 | NM_146748.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P519632 | AF121983.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P519632 | NM_146748.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P519632 | AF121983.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P135019 | - | - |
| Whole Mouse Genome G4122A | A_51_P519632 | NM_146748.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P519632 | AF121983.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P519632 | NM_146748.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P519632 | AF121983.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P105235 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P135019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389110 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05973963 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389128 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389179 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389199 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389142 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389181 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389131 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389129 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06153124 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389198 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389192 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06394061 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06332042 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06281538 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389154 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06091894 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389136 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389124 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389178 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389166 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06281541 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389171 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06153126 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06091883 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06281555 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389119 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389150 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389157 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389173 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389151 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389177 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389110 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389117 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389164 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389121 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389112 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389155 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389163 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389169 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389144 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389126 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389170 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389140 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06281536 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06091895 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389156 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389108 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389165 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389188 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389186 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389115 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389194 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389125 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389168 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389187 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10555888 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10555887 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700012910.mm.1 | TC0700001532.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700012910.mm.1 | NM_146748.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700012910.mm.1 | NM_146748.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2850538 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002850538 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2688387 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2688387 | - | - |
| mouse-6 v1.1 (Illumina) | scl32251.1.399_46-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2850538 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl32251.1.399_46-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2850538 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2688387 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2688387 | - | - |

Olfr661
Genomic Map