Assay Details
| Gene Symbol: Insc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC026405.1 | - | 1 - 2 | 128 | 4211657 | 76 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC026405.1 transcripts.
Set Membership: |
|

Insc
Genomic Map