Assay Details
| Gene Symbol: Olfr531 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146953.1 | NP_667164.1 | 1 | 470 | 75 | |
| GenBank mRNA | BC104052.1 | - | 1 | 470 | 40040241 | 75 |
| BC104053.1 | - | 1 | 470 | 40040242 | 75 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Olfr531
Genomic Map