Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700019820.mm.1 |
NM_001033493.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019820.mm.1 |
NM_001033493.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019820.mm.1 |
TC1700002117.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700019818.mm.1 |
NM_001033493.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019818.mm.1 |
NM_001033493.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019818.mm.1 |
TC1700002117.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700019819.mm.1 |
NM_001033493.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019819.mm.1 |
NM_001033493.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700019819.mm.1 |
TC1700002117.mm.1 |
MTA TC |