Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700033622.mm.1 |
NM_054103.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700033622.mm.1 |
NM_054103.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700033622.mm.1 |
TC0700004073.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700033619.mm.1 |
NM_054103.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700033619.mm.1 |
NM_054103.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700033619.mm.1 |
TC0700004073.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700033621.mm.1 |
NM_054103.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700033621.mm.1 |
NM_054103.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700033621.mm.1 |
TC0700004073.mm.1 |
MTA TC |