Assay Details
| Gene Symbol: Hsd17b7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010476.3 | NP_034606.3 | 4 - 5 | 520 | 70 | |
| GenBank mRNA | AK028380.1 | - | 4 - 5 | 476 | 70 | |
| AK050211.1 | - | 4 - 5 | 477 | 70 | ||
| AK077914.1 | - | 4 - 5 | 518 | 70 | ||
| BC011464.1 | - | 4 - 5 | 484 | 70 | ||
| Y15733.1 | - | 4 - 5 | 510 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hsd17b7
Genomic Map