Assay Details
| Gene Symbol: 1700015E13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039593.1 | NP_001034682.1 | 1 - 2 | 124 | 81 | |
| GenBank mRNA | AK005990.1 | - | 1 - 2 | 183 | 81 | |
| BC099563.1 | - | 1 - 2 | 124 | 30916762 | 81 | |
| BC132313.1 | - | 1 - 2 | 123 | 40130590 | 81 | |
| BC132315.1 | - | 1 - 2 | 123 | 40130592 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P276614 | NM_001039593.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P276614 | AK005990.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P276614 | BC099563.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P276614 | NM_001039593.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P276614 | AK005990.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P276614 | BC099563.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 265375 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P276614 | NM_001039593.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P276614 | AK005990.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P276614 | BC099563.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P276614 | NM_001039593.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P276614 | AK005990.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P276614 | BC099563.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20821658 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20821692 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20821670 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20821684 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20821663 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20821648 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20821682 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20821695 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20821656 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20821694 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20821672 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20821680 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20821691 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20821675 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20821652 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20821668 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20821665 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20821688 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20821679 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20821660 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20821659 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20821662 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20821678 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20821650 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20821676 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20821673 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20821654 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20821686 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20821666 | - | - |
| Whole Mouse Genome G4122A | A_52_P276614 | NM_001039593.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P276614 | AK005990.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P276614 | BC099563.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P276614 | NM_001039593.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P276614 | AK005990.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P276614 | BC099563.1 | mouse_transcriptome |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00569638 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00392584 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00392579 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00392581 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00392587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20821711 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00538215 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20821708 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20821625 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20821682 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20821634 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20821695 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20821684 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20821717 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20821632 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00771186 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20821688 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20821691 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20821618 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20821611 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20821711 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20821662 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20821658 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20821675 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20821702 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20821609 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20821706 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20821718 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20821619 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20821700 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00566254 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20821685 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20821677 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20821652 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20821686 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20821676 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20821680 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00771190 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20821714 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20821663 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20821622 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20821643 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20821624 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20821629 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20821710 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00392587 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20821665 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20821692 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20821679 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20821633 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20821672 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20821654 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00181538 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00082694 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20821621 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20821703 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00289546 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00113840 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20821694 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20821620 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00048101 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20821659 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00187478 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20821636 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00082692 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00538217 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00538200 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00205342 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20821631 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00289536 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20821641 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20821615 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20821670 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20821720 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00771188 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20821721 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20821673 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20821647 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20821674 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10351486 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10351484 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1437384_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10351482 | - | - |
| Mouse Genome 430 2.0 Array | 1437384_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1437384_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 76925_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 76925_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100012847.mm.1 | NM_001039593.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100012847.mm.1 | NM_001039593.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100012847.mm.1 | TC0100001569.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100012846.mm.1 | NM_001039593.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100012846.mm.1 | NM_001039593.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100012846.mm.1 | TC0100001569.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100012845.mm.1 | NM_001039593.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100012845.mm.1 | NM_001039593.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100012845.mm.1 | TC0100001569.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1437384_PM_at | - | - |
| Mouse Expression Set 430 | 1437384_at | - | - |
| Murine Genome U74v2 Set | 169089_i_at | - | - |
| Murine Genome U74v2 Set | 169089_PM_i_at | - | - |
| Murine Genome U74v2 Set | 170428_PM_i_at | - | - |
| Murine Genome U74v2 Set | 170428_i_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1437384_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1437384_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 624493 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2634508 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2634508 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2634503 | - | - |

1700015E13Rik
Genomic Map