Assay Details
| Gene Symbol: Rgs5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009063.3 | NP_033089.2 | 1 - 2 | 126 | 65 | |
| GenBank mRNA | AK004165.1 | - | 1 - 2 | 127 | 65 | |
| AK028765.1 | - | 128 | 65 | |||
| AK030300.1 | - | 127 | 65 | |||
| AK044096.1 | - | 1 - 2 | 127 | 65 | ||
| AK054098.1 | - | 1 - 2 | 127 | 65 | ||
| BC037683.1 | - | 1 - 2 | 113 | 3962083 | 65 | |
| U67188.1 | - | 83 | 65 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Rgs5
Genomic Map