Assay Details
| Gene Symbol: Uap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK036243.1 | - | 4 - 5 | 842 | 97 | |
| AK047566.1 | - | 4 - 5 | 907 | 97 | ||
| AK049519.1 | - | 4 - 5 | 896 | 97 | ||
| AK081690.1 | - | 4 - 5 | 867 | 97 | ||
| AK147139.1 | - | 4 - 5 | 842 | 97 | ||
| AK163314.1 | - | 4 - 5 | 741 | 97 | ||
| AK171925.1 | - | 4 - 5 | 97 | |||
| BC016406.1 | - | 4 - 5 | 4489112 | 97 | ||
| BC017547.1 | - | 4 - 5 | 4920702 | 97 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Uap1
Genomic Map