Assay Details
| Gene Symbol: Uap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK036243.1 | - | 6 - 7 | 1209 | 77 | |
| AK047566.1 | - | 6 - 7 | 1274 | 77 | ||
| AK049519.1 | - | 6 - 7 | 1263 | 77 | ||
| AK081690.1 | - | 6 - 7 | 1234 | 77 | ||
| AK147139.1 | - | 6 - 7 | 1209 | 77 | ||
| AK163314.1 | - | 6 - 7 | 1108 | 77 | ||
| AK171925.1 | - | 6 - 7 | 1106 | 77 | ||
| AK178045.1 | - | 1 - 2 | 143 | 77 | ||
| BC016406.1 | - | 6 - 7 | 1196 | 4489112 | 77 | |
| BC017547.1 | - | 6 - 7 | 1085 | 4920702 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Uap1
Genomic Map