Assay Details
Gene Symbol: Mtm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019926.3 | NP_064310.2 | 1 - 2 | 68 | 126 | |
GenBank mRNA | AF073996.1 | - | 1 - 2 | 69 | 126 | |
AK166759.1 | - | 1 - 2 | 66 | 126 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF073996.1, NM_019926.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
