Assay Details
Gene Symbol: Mtm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164190.1 | NP_001157662.1 | 3 - 4 | 324 | 167 | |
NM_001164191.1 | NP_001157663.1 | 3 - 4 | 215 | 167 | ||
NM_001164192.1 | NP_001157664.1 | 3 - 4 | 215 | 167 | ||
NM_001164193.1 | NP_001157665.1 | 3 - 4 | 215 | 167 | ||
NM_019926.3 | NP_064310.2 | 3 - 4 | 204 | 167 | ||
GenBank mRNA | AF073996.1 | - | 3 - 4 | 205 | 167 | |
AK016445.1 | - | 3 - 4 | 325 | 167 | ||
AK031794.1 | - | 3 - 4 | 186 | 167 | ||
AK032870.1 | - | 3 - 4 | 168 | 167 | ||
AK149997.1 | - | 3 - 4 | 217 | 167 | ||
AK166452.1 | - | 3 - 4 | 183 | 167 | ||
AK171475.1 | - | 3 - 4 | 193 | 167 | ||
BC051022.1 | - | 3 - 4 | 173 | 167 | ||
BC090984.1 | - | 3 - 4 | 182 | 30246206 | 167 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
