Assay Details
| Gene Symbol: Bhmt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022884.2 | NP_075022.2 | 5 - 6 | 629 | 57 | |
| GenBank mRNA | AF257474.1 | - | 5 - 6 | 615 | 57 | |
| AK090308.1 | - | 5 - 6 | 607 | 57 | ||
| AK145901.1 | - | 5 - 6 | 630 | 57 | ||
| BC013515.1 | - | 5 - 6 | 626 | 4235455 | 57 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bhmt2
Genomic Map