Assay Details
| Gene Symbol: Dmgdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028772.3 | NP_083048.1 | 11 - 12 | 1798 | 115 | |
| GenBank mRNA | AK004755.1 | - | 11 - 12 | 1799 | 115 | |
| BC024126.1 | - | 7 - 8 | 1081 | 115 | ||
| BC089599.1 | - | 11 - 12 | 1801 | 30118948 | 115 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dmgdh
Genomic Map