Assay Details
| Gene Symbol: Hexb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010422.2 | NP_034552.1 | 7 - 8 | 932 | 99 | |
| GenBank mRNA | AK151149.1 | - | 7 - 8 | 869 | 99 | |
| AK159134.1 | - | 7 - 8 | 885 | 99 | ||
| AK159178.1 | - | 7 - 8 | 906 | 99 | ||
| AK160560.1 | - | 7 - 8 | 901 | 99 | ||
| AK168825.1 | - | 7 - 8 | 906 | 99 | ||
| U07633.1 | - | 7 - 8 | 904 | 99 | ||
| Y00964.1 | - | 7 - 8 | 845 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hexb
Genomic Map