Assay Details
Gene Symbol: Col4a3bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164222.1 | NP_001157694.1 | 13 - 14 | 1839 | 151 | |
NM_023420.2 | NP_075909.1 | 14 - 15 | 1917 | 151 | ||
GenBank mRNA | AF232932.1 | - | 14 - 15 | 1931 | 151 | |
AF232934.1 | - | 13 - 14 | 1853 | 151 | ||
AK012989.1 | - | 13 - 14 | 1820 | 151 | ||
AK018103.1 | - | 2 - 3 | 208 | 151 | ||
AK030305.1 | - | 14 - 15 | 1747 | 151 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
