Assay Details
| Gene Symbol: Col4a3bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164222.1 | NP_001157694.1 | 6 - 7 | 1109 | 149 | |
| NM_023420.2 | NP_075909.1 | 6 - 7 | 1109 | 149 | ||
| GenBank mRNA | AF232932.1 | - | 6 - 7 | 1123 | 149 | |
| AF232934.1 | - | 6 - 7 | 1123 | 149 | ||
| AK012989.1 | - | 6 - 7 | 1090 | 149 | ||
| AK030305.1 | - | 7 - 8 | 1017 | 149 | ||
| AK038803.1 | - | 6 - 7 | 1090 | 149 | ||
| AK047100.1 | - | 6 - 7 | 1106 | 149 | ||
| AK080958.1 | - | 6 - 7 | 1107 | 149 | ||
| AK163624.1 | - | 6 - 7 | 1099 | 149 | ||
| BC016197.1 | - | 6 - 7 | 1072 | 4502118 | 149 | |
More Information
Updated Mapping Information
Set Membership: |
|

Col4a3bp
Genomic Map