Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1300006931.mm.1 |
NM_001164222.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006931.mm.1 |
NM_001164222.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006931.mm.1 |
NM_023420.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006931.mm.1 |
NM_023420.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006931.mm.1 |
TC1300001089.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300006933.mm.1 |
NM_001164222.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006933.mm.1 |
NM_001164222.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006933.mm.1 |
NM_023420.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006933.mm.1 |
NM_023420.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006933.mm.1 |
TC1300001089.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300006934.mm.1 |
NM_001164222.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006934.mm.1 |
NM_001164222.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006934.mm.1 |
NM_023420.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006934.mm.1 |
NM_023420.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300006934.mm.1 |
TC1300001089.mm.1 |
MTA TC |